missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ATG5 Recombinant Protein Antigen

Product Code. 18270198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
Product Code. Quantity unitSize
18270198 100 μg 100µL
1 options
This item is not returnable. View return policy

Product Code. 18270198

Brand: Novus Biologicals™ NBP288936PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9474
Protein Family ATG5
Purity >80%, by SDS-PAGE
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS, 1 M Urea, pH 7.4.
Gene Symbol ATG5
Quantity 100 μg
Research Category Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-55080. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.