missing translation for 'onlineSavingsMsg'
Learn More

uvex Athletic All-Round Assembly Gloves

Product Code. 17230341 Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17230341

Brand: uvex 6002808

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, dirt-resistant gloves are constructed from a blend of polyamide and elastane, with an NBR foam coating for enhanced grip capabilities.

  • Slim-fit provides tactile feedback when assembling smaller workpieces
  • NBR foam coating is abrasion resistant and enhances grip in dry or slightly damp environments
  • Dirt-resistant shell is ideal for mechanical tasks
  • Certified by OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material NBR Foam
Color Gray, Black
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Elastane, Polyamide
Product Type Mechanical Protection Gloves
Size 8
EN388 Standard 4122X
For Use With (Application) Assembly, Packaging, Precision Work, and Transportation
Gender Unisex
Glove Cuff Style Knit
Series Athletic All-Round
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.