missing translation for 'onlineSavingsMsg'
Learn More

uvex Athletic Lite Assembly Gloves

Product Code. p-7230481 Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17727559

Brand: uvex 6002706

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are constructed from a combination of polyamide and elastane, with a nitrile rubber microfoam coating for enhanced grip in dry or slightly damp applications.

  • Slim, ergonomic fit allows for precision work with high levels of user comfort
  • Fine-structured, open-pored microfoam coating enhances grip capabilities
  • Breathable, matte coating reduces excessive perspiration
  • Ideal for precision assembly work, maintenance, inspection, and sorting
  • Certified by OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Coating Material NBR MicroFoam
Color Blue, Black
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 6
EN388 Standard 4132X
For Use With (Application) Inspection, Precision Assembly Work, Repair Work, and Sorting
Gender Unisex
Series Athletic Lite
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.