missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atrazine-d5, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Atrazine-d5 |
| CAS | 163165-75-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 2H5 H9 Cl N5 |
| Quantity | 10 mg |
| Synonym | Atrazine D5 (ethyl D5), 2-Chloro-4-pentadeuteroethylamino-6-isopropylamine-1,3,5-triazine, Atrazine-d5 |
| InChI Formula | InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)/i1D3,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC(C)C)n1 |
| IUPAC Name | 6-chloro-4-N-(1,1,2,2,2-pentadeuterioethyl)-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 220.71 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction