Learn More
Invitrogen™ Attune™ NxT Custom Filter Holder Kit
Allows users of the Attune™ NxT acoustic focusing cytometer to make their own custom emission or dichroic filters in order to customize the optical configuration of the instrument
Brand: Invitrogen™ A27784
Additional Details : Weight : 1.00000kg
Description
- The Attune™ NxT Custom Filter Holder Kit allows users of the Attune™ NxT acoustic focusing cytometer to make their own custom emission or dichroic filters in order to customize the optical configuration of the instrument.
- Each kit contains sufficient parts to make 2 emission filters and 2 dichroic filters.
- The kit includes instructions for attaching the filter to the filter blade, but does not include the glass filter.
Specifications
Attune NxT Acoustic Focusing Cytometer | |
Custom Filter Holder Kit | |
1 Kit |
2 dichroic filter blades 2 emission filter blades 4 clips Instruction card |
|
Approved for shipment at Room Temperature or on Wet Ice | |
Attune™ |