Learn More
Attune™ NxT Dichroic Filter, 690DLP
Attune NxT Dichroic Filter, 690DLP
Brand: Invitrogen 100022773
Additional Details : Weight : 0.10000kg
Description
This is a replacement 690 DLP dichroic filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.Specifications
Attune Acoustic Focusing Cytometer | |
Room temperature | |
Attune™ NxT Dichroic Filter | |
1 Each |
For Research Use Only. Not for use in diagnostic procedures.