Learn More
Attune™ NxT Emission Filter, 620/15BP
Attune NxT Emission Filter, 620/15BP
Brand:  Invitrogen 100022762
Additional Details : Weight : 0.10000kg
Product Code. 15350647
  • 187.00€ / Each
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
This is a replacement 620/15 bandpass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Specifications
Specifications
Attune Acoustic Focusing Cytometer
Room temperature
Attune™ NxT Emission Filter
1 Each