Learn More
Attune™ NxT OD2-488/10 Filter
This is a replacement OD2-488/10 filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Brand:  Invitrogen 100022768
Additional Details : Weight : 0.10000kg
Product Code. 15310657
  • 355.00€ / Each
Estimated Shipment: 06-05-2024
to see stock.
Description
Description
  • Replacement filter
  • Used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer
Specifications
Specifications
Attune Acoustic Focusing Cytometer
Store at room temperature.
Filter
Attune™