Learn More
3M™ Aura™ Particulate Respirator, Series 9400
Click to view available options
Filter Type:
FFP2
FFP3
Unit Size:
Pack of 10
Description
- Provide reliable and comfortable protection from a range of airborne disposable hazards
- Minimize contamination risks to food or pharmaceutical production processes
- Made without staples or small detachable parts
- Cool Flow™ exhalation valve option provides effective removal of heat and moisture build up for a more comfortable wear
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 149:2001 + A1:2009 |
| Color | Blue |
| Filter Type | FFP3 |
| Sterility | Non-sterile |
| FDA Cleared Surgical Mask | No |