Learn More
Abeba™ Autoclavable Clogs 9630
Antistatic and autoclavable at 121°C. High slip resistance.
54.30€ - 57.56€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E SRA |
---|---|
Color | Orange |
Style | 9630 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15683268
|
Abeba™
9630/36 |
36 |
57.56€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15693268
|
Abeba™
9630/37 |
37 |
57.56€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15603278
|
Abeba™
9630/38 |
38 |
54.30€
1 pair |
In Stock
Log in to see stock. |
|||||
15613278
|
Abeba™
9630/39 |
39 |
57.56€
1 pair |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15623278
|
Abeba™
9630/40 |
40 |
57.56€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15633278
|
Abeba™
9630/41 |
41 |
57.56€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15643278
|
Abeba™
9630/42 |
42 |
57.56€
1 pair |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15653278
|
Abeba™
9630/43 |
43 |
57.56€
1 pair |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15663278
|
Abeba™
9630/44 |
44 |
57.56€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15673278
|
Abeba™
9630/45 |
45 |
57.56€
1 pair |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15683278
|
Abeba™
9630/46 |
46 |
57.56€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E SRA
- Autoclavable, sterilisable special material
- Orange
- Antistatic
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
CE EN ISO 20347:2012 OB A, E SRA | |
Orange | |
9630 |