Learn More
Poulten Graf™ Autoclavable Glass Syringe
Able to be autoclaved at 134°C. Poulten Graf™ Autoclavable Glass Syringes come with an interchangeable piston.
6.75€ - 80.45€
Product Code | Brand | Volume (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Description | Price | Quantity & Availability | ||||
10579032
|
Poulten Graf™
7 14051 |
100 mL | Luer-Lock Tip |
80.45€
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10645411
|
Poulten Graf™
7 10251 |
100 mL | Luer Tip |
63.90€
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10373421
|
Poulten Graf™
7 10221 |
1 mL | Luer Tip |
6.75€
Each |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10613551
|
Poulten Graf™
7 10241 |
20 mL | Luer Tip |
13.90€
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10723491
|
Poulten Graf™
7 10244 |
30 mL | Luer Tip |
19.25€
Each |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10713491
|
Poulten Graf™
7 10229 |
3 mL | Luer Tip |
6.90€
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Complies with CE EN 46001 standards
- Constructed from Durobax™ glass
- With permanent amber graduation
- Acid and alkali resistant
Specifications
Autoclavable | |
Syringe | |
1/Ea. |