Learn More
Cole-Parmer™ Parker Hannifin™ PFA Threaded Adapter Compression Fitting
Click to view available options
For Use With:
1/2 tubing O.D. x 1/2 in. NPT(M) Thread
1/4 tubing O.D. x 1/4 in. NPT(F) Thread
1/4 tubing O.D. x 1/8 in. NPT(M) Thread
Thread Size:
1/2 in. NPT(M)
1/4 in. NPT(F)
1/8 in. NPT(M)
Unit Size:
Each
Beschreibung
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and use the nut to compress down to form a secure seal
- Superior chemical resistance with durable PFA
- Available in male and female varieties
- Sterilization through autoclave or ethylene oxide (ETO)
Spezifikation
Spezifikation
| Autoclavable | Autoclavable |
| Material | PFA |
| For Use With | 1/2 tubing O.D. x 1/2 in. NPT(M) Thread |
| Packaging Quantity | 1 Each |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Thread Size | 1/2 in. NPT(M) |
| Product Type | Compression Fitting |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur