Learn More
Keystone Cleanroom Products Autoclavable Tyvek™ Stopper Bags with Heat Seal Closure
GREENER_CHOICE
Product Code. 17821721 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Length (English):
14 in.
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) unitSize
17821721 14 in. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17821721 Supplier Keystone Cleanroom Products Supplier No. 1414SBSD
Only null left
Add to basket
Add to basket
These Tyvek™ Stopper Bags will not rip or puncture. Includes heat seal closure and double-welded factory seals.
The medical Grade 1073B Tyvek™ material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
  • Manufactured and packaged in an ISO certified cleanroom
  • Double Welded Factory Seals
  • Autoclavable at up to 125°C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Heat seals closed
  • Integrated steam sterilization indicator
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Heat Seal
Color Clear, White
Disposable Single-use
For Use With (Application) Steam, EtO, Gamma or VHP Sterilization
For Use With (Equipment) Pharmaceutical Equipment
Includes Steam Indicator Dot
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Quantity 500 Pk.
Sterilization Method Steam
Temperature Range (English) <250°F
Temperature Range (Metric) <125°C
Dimensions (L x W) 14 x 14 in.
Length (English) 14 in.
Width (English) 14 in.
Product Type Autoclave Bag
Length (Metric) 35.6 cm
Width (Metric) 35.6 cm