Learn More
Microspec™ Autoclave Bags
Product Code. 15722135
Click to view available options
Length (Metric):
302 mm
632 mm
633 mm
Outer Packaging:
Bag
EcoPak
Quantity:
200
1200
1500
Unit Size:
Pack of 1500
Pack of 200
Pack of 500
Product Code. 15722135
Brand: Microspec™ H1
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Designed for contaminated waste disposal in autoclaves or incinerators.
  • Clear bags imprinted with blue biohazard labeling designed for safe disposal
  • Constructed of thick polypropylene
  • Allows for repeated autoclaving
  • Strong side seams for securely holding items
  • Front write-on panel
Specifications
Content And Storage Room Temperature
Autoclavable Autoclavable
Color Clear with Blue Print
Disposable Single-use
Material Polypropylene
Sterility Non-sterile
Thickness (Metric) 50 μm
Film Description Polypropylene
No. per Case 200
Outer Packaging EcoPak
Quantity 1500
Size Medium
Thickness (English) 2 mil
Length (Metric) 632 mm
Width (English) 16.26 in.
Width (Metric) 413 mm
Height (Metric) 413 mm
Weight (English) 62.733 lb.
Weight (Metric) 28.455 kg
Product Type Autoclave Bags