Learn More
Cytiva AutoSeq G-50 Columns
Use for rapid removal of unincorporated fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing. Cytiva Life Sciences™ illustra™ AutoSeq G-50 Columns provide effective purification essential for high-quality sequencing results.
Brand: Cytiva 27-5340-01
Additional Details : CAS : 9048-46-8 Weight : 0.21100kg
Description
- Columns are prepacked with Sephadex G-50 DNA Grade F and pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
- Columns that are ready to use enable sample application to collection of purified product in less than 4 min.
- Can be used to clean up reactions prepared with the BigDye Terminator v3.1 Cycle Sequencing Kit from Applied Biosystems
- Typical sample volume 12 to 25 μl
- Suitable for purification of DNA greater than 20 bases in length
- Optimized for automated fluorescent sequencing
Specifications
Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing | |
Column | |
50 Columns |