missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Auxinole
GREENER_CHOICE

Product Code. p-200009226
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000437

Brand: Tocris Bioscience™ 7894/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Auxin antagonist of TIR1/AFB receptors; also OsTIR1 inhibitor in auxin-inducible degron (AID) system

Auxinole is an auxin antagonist of TIR1/AFB receptors. Binds to TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and inhibit auxin-responsive gene expression. Auxinole competitively inhibits various auxin responses in planta. Auxinole also inhibits OsTIR1 that suppresses leaky degradation of degron-fused proteins. Can be used as a control compound with 5-Ph-IAA (Cat. No. 7392) in the AID2 degron system to switch off degradation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material α-[2-(2,4-Dimethylphenyl)-2-oxoethyl]-1H-indole-3-acetic acid
CAS 86445-22-9
Quantity 10 mg
Target Molecular Glues
Molecular Formula C20H19NO3
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.