missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ AVE 1625
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AVE 1625 is a potent and selective CB1 receptor antagonist (IC50 values are 25 and 10 nM for human and rat CB1R, respectively). Exhibits >400-fold selectivity for CB1 over CB2 receptors. Attenuates Olanzapine (Cat. No. 4349) induced weight gain and food intake in rats with no effect on energy expenditure or motility. Also improves cognitive function in rodent schizophrenia models and ameliorates extrapyramidal side-effects of antipsychotics.
Specifications
Specifications
| Synonym | Drinabant |
| Quantity | 10 mg |
| Molecular Formula | C23H20Cl2F2N2O2S |
| Formula Weight | 497.38 |
| Product Type | Antagonist |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction