missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Avian Influenza A H7N9 Hemagglutinin ELISA Kit (Colorimetric)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Assay Range | 15.6-1000 pg/mL |
| Assay Sensitivity | 3 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Recombinant Protein |
| Specificity | Recognizes both recombinant and natural Avian Influenza A H7N9 Hemagglutinin. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Quantity | 1 Kit |
| Research Discipline | Infections (Virus Bacteria and Parasites) |
| Storage Requirements | Storage is content dependent. |
| Synonym | H7N9HA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction