missing translation for 'onlineSavingsMsg'
Learn More

AW Standard Super-Cel™ NF Diatomaceous Earth, Honeywell Fluka™

Catalog Number p-7120205
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
91053-39-3
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
This item is not returnable. View return policy
91053-39-3
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O

Suitable for most filtrations, derived from diatomaceous earth. Super Cel is a registered trademark of Imerys Minerals California, Inc.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 250 g
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Aw Standard Super-Cel™ Nf
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'