Learn More
Carl Zeiss™ AxioVision Rel. 4.8.2 Software
Control high-sensitivity microscopes, and capture, process, annotate, and catalog images with this software download. Carl Zeiss™ AxioVision Rel. 4.8.2 Software helps you perform complex and minute tasks simply and intuitively.
Brand:  Carl Zeiss™ 410130-0600-000
Additional Details : Weight : 0.80000kg
Product Code. 11875113
  • 3439.70€ / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Images can be acquired using any common camera with TWAIN driver
  • Open and compare up to 21 different image file formats including multidimensional ZVI-images
  • Adjust display curve, brightness, contrast and color balance
  • Acquire length, contour and angle of microscopical structures. Export the results to other programs
  • Apply comments and scale bars to your image
  • Print directly from AxioVision LE or export your image to various other file formats
Specifications
Specifications
AxioVision Rel. 4.8.2 Software
Light microscopes, PC
1 Ea.