Learn More
Invitrogen™ Human AXL, His Tag Recombinant Protein
The purified recombinant protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ assay technologies to identify novel kinase inhibitors. AXL is also available in our SelectScreen™ Kinase Profiling Service.
634.00€ - 1796.00€
Specifications
Accession Number | NP_068713.2 |
---|---|
For Use With (Application) | Kinase Assay |
KinaseFamily | AXL Kinase Family |
Protein Length | 473 - end |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15647888
|
Invitrogen™
A31515 |
10μg |
634.00€
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15657888
|
Invitrogen™
A31516 |
100μg |
1796.00€
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AXL is a chronic myelogenous leukemia-associated oncogene that is also associated with colon cancer and melanoma. The AXL protein is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6, and is involved in the stimulation of cell proliferation.
- Validated for use with the LanthaScreen & Z'-LYTE assay technology
- Available in multiple pack sizes for convenient use
- Also available in our SelectSceen Kinase Profiling Service
Specifications
NP_068713.2 | |
AXL Kinase Family | |
Human | |
His Tag |
Kinase Assay | |
473 - end | |
Recombinant | |
Insect |