Learn More
Axygen™ 20 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
For use with Agilent/Velocity11 automation systems
Brand:  Axygen™ VT-20-R
Additional Details : Weight : 9.50000kg
 View more versions of this product
Product Code. 12595557
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Available in nonsterile or sterile
Specifications
Specifications
20 μL
Non-sterile
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Rack
SBS Rack
Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Non-Filtered
Clear
DNase, Rnase Free
w/out Bottom Label
Automation, Round
50 Racks/Cs.