Learn More
Axygen™ PerkinElmer™/Packard Robotic Tips: Maxymum Recovery
Ideal for MiniTrak, PlateTrak and Evolution P-3
Brand:  Axygen™ PK-200-L-R
Additional Details : Weight : 9.63000kg
 View more versions of this product
Product Code. 12732896
  • 442.00€ / Pack of 4800
Estimated Shipment: 06-10-2021
Add to basket
Description
Description
Robotic tips uses no mold-releasing agents. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.
  • Tip volume: 20 to 200μL
  • Tip color: clear
  • Rack color: green
Specifications
Specifications
200 μL
Non-sterile
Green
PP (Polypropylene)
DNase, Rnase Free
PerkinElmer™ Janus™, PerkinElmer™ Evolution P3™, PerkinElmer™ MiniTrak™, PerkinElmer™ PlateTrak™, PerkinElmer™ MDT-96 P235, PerkinElmer™ MDT-96 P50, PerkinElmer™ MDT-384 30μL
Rack
SBS Rack
Non-Filtered
Clear
Round
ANSI
Maxymum Recovery™, non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
PerkinElmer™ Janus™, PerkinElmer™ Evolution P3™, PerkinElmer™ MiniTrak™, PerkinElmer™ PlateTrak™, PerkinElmer™ MDT-96 P235, PerkinElmer™ MDT-96 P50, PerkinElmer™ MDT-384 30μL
Automation
50 Racks/Cs.