Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
GREENER_CHOICE
Product Code. 12637765 Shop All Axygen Scientific Products
Click to view available options
Product Type:
Heat Sealing Film for Compound Storage
Heat Sealing Film for qPCR
Peelable Heat Sealing Film
Peelable Heat Sealing Film for qPCR
Pierceable Aluminum Heat Sealing Film
Unit Size:
Pack of 500
Product Code. 12637765
Brand: Axygen™ HS300
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Plate Sealing
For Use With (Equipment) PlateMax Plate Sealer
Material Polyester, Aluminum
No. per Case 500/Cs.
No. per Pack 100/Pk.
Product Type Pierceable Aluminum Heat Sealing Film