Learn More
Axygen™ Polypropylene Half Skirt 96-Well PCR Microplates
Brand:  Axygen PCR-96M2-HS-BK
Additional Details : Weight : 1.40000kg
Product Code. 12719478
  • 347.00€ / Pack of 50
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Specifications
Yes
8 x 12
Polypropylene
Yes
10/Pk.
Standard
Black
96
4000 x g
Yes