Learn More
Axygen™ Screw Cap Tubes without Caps: Conical
Product Code. 11364215 Shop All Axygen Scientific Products
Click to view available options
Color:
Amber
Clear
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Bottom Shape:
Conical
Elangated Conical
Unit Size:
Pack of 4000
Pack of 500
Product Code. 11364215
Brand: Axygen™ ST050
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Conical tubes
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
  • Tube volumes from 0.5 to 2.0mL
  • Choose clear or amber
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Capacity (Metric) 0.5 mL
Color Clear
Material Polypropylene
Packaging 500 Tubes/Pk., 8 Packs/Cs.
Sterility Non-sterile
Graduated No
Closure Type Not Included
Bottom Shape Elangated Conical
Max. RCF 20,000 x g