missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL

Clear, autoclavable tips; color-coded racks.

Brand:  Axygen™ ZT-100-R-S

Additional Details : Weight : 10.43000kg

 View more versions of this product

Product Code. 12796669

  • 325.00€ / Pack of 4800


Estimated Shipment: 13-08-2021
Add to basket

Description

Description

  • Tip format: 96 tip and 384 tip
  • Tip volume range: 25 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
  • Rack format: Non-ANSI (96) and ANSI/SLAS (384)
  • Rack material: Polypropylene

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

Specifications

Specifications

100 μL
Sterile
Gray
Round
Non-ANSI
Tecan™ Freedom EVO™, Tecan™ TeMo, Tecan™ Aquarius MCA96 head, Zymark™/Caliper™ RapidPlate™, SciClone™, SciClone™ HV96
Automation
50 Racks/Cs.
Non-Filtered
Clear
Autoclavable
PP (Polypropylene)
Tecan™ Freedom EVO™, Tecan™ TeMo, Tecan™ Aquarius MCA96 head, Zymark™/Caliper™ RapidPlate™, SciClone™, SciClone™ HV96, CAS-4200-1ch. 20-150μL/sec. head
Rack
Non-SBS Rack
Videos
SDS
Documents
Certificates