Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 50μL
Brand:  Axygen ZT-50-R
Additional Details : Weight : 10.90000kg
 View more versions of this product
Product Code. 12706689
  • 286.00€ / Pack of 4800
Estimated Shipment: 16-08-2021
Add to basket
Description
Description
  • High throughput screening
  • Volume: 50µL
  • Rack Color: Light Green
  • Color: Clear
  • Designed for use with Zymark™ RapidPlate™, Zymark™ SciClone, Tecan™ Te-MO™, Tecan™ Genmate™ and Tecan™ Aquarius™ automated liquid handling systems
  • Certified RNase, DNase and Pyrogen-Free
  • Format: 96 well
Specifications
Specifications
50 μL
Non-sterile
Light Green
Round
Non-ANSI
Tecan™ Freedom EVO™, Tecan™ TeMo, Tecan™ Aquarius MCA96 head, Zymark™/Caliper™ RapidPlate™, SciClone™, SciClone™ HV96, CAS-4200-1ch. 20-150μL/sec. head
Rack
Non-filtered
Clear
Autoclavable
PP (Polypropylene)
Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
Tecan™ Freedom EVO™, Tecan™ TeMo, Tecan™ Aquarius MCA96 head, Zymark™/Caliper™ RapidPlate™, SciClone™, SciClone™ HV96
Automation