missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ PFKFB3 67, >98%

Description
Potent and selective PFKFB3 inhibitor (IC50 values are 11, 159 and 1130 nM for PFKFB3, PFKFB2 and PFKFB1 respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Formula Weight | 455.51 |
| Purity | >98% |
| Molecular Weight (g/mol) | 455.518 |
| Chemical Name or Material | (2S)-N-[4-[[3-Cyano-1-[(3,5-dimethyl-4-isoxazolyl)methyl]-1H-indol-5-yl]oxy]phenyl]-2-pyrrolidinecar |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?