missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZD 1283
GREENER_CHOICE

Product Code. 18768219
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18768219

Brand: Tocris Bioscience™ 6085/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity P2Y12 antagonist

AZD 1283 is a potent and high affinity P2Y12 antagonist (IC50 = 11 nM in radioligand competition binding assay, and 25 nM in a GTPγS binding assay). Causes a dose-dependent increase in blood flow and inhibits ADP-induced platelet aggregation in vivo. Antithrombotic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 5-cyano-2-methyl-6-[4-[[[(phenylmethyl)sulfonyl]amino]carbonyl]-1-piperidinyl]-3-pyridinecarboxylate
CAS 919351-41-0
Synonym Ethyl 5-cyano-2-methyl-6-[4-[[[(phenylmethyl)sulfonyl]amino]carbonyl]-1-piperidinyl]-3-pyridinecarboxylate
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Target P2Y Receptor Antagonists
Molecular Formula C23H26N4O5S
Formula Weight 470.54
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity 98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.