missing translation for 'onlineSavingsMsg'
Learn More

Azidoethanol, TRC

Product Code. 30004836
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004836 1 g 1g
30136239 10 g 10g
30136594 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004836 Supplier Toronto Research Chemicals Supplier No. TRCA8485601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1517-05-1
Formula Weight 87.0433
InChI Formula InChI=1S/C2H5N3O/c3-5-4-1-2-6/h6H,1-2H2
IUPAC Name 2-azidoethanol
Molecular Formula C2 H5 N3 O
Molecular Weight (g/mol) 87.08
Percent Purity >95%
SMILES OCCN=[N+]=[N-]
Synonym Ethanol, 2-azido-, -Azido-2-hydroxyethane, 2-Azido-1-ethanol, 2-Azidoethanol, 2-Hydroxyethyl azide, Azidoethanol, 2-Azidoethanol
Chemical Name or Material 2-Azidoethanol
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.