Learn More
Azlon™ Acrylic Single-Bore Class B Buret
Product Code. 12044637
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Product Code. 12044637
Brand: Azlon BDH112T
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Suitable for left or right handed use. The Azlon™ Acrylic Single Bore Class B Buret is great for educational laboratories, especially where breakages often occur.
  • Acrylic construction for strength and transparency
  • Graduations are screen printed in black to ISO 385 Class B accuracy
  • Suitable for use in educational laboratories especially where breakages often occur
  • Unaffected by most dilute aqueous solutions, however unsuitable for use with organic solvents
  • Turning the stopcock through 90° makes it suitable for left or right handed use
  • Stopcocks can be removed for easy cleaning
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25 mL
Material Acrylic
Graduations 0.1 mL
For Use With (Application) Educational Laboratories Especially where Breakages Often Occur
Includes PMP/Polyethylene Stopcocks
Stopcock Material PMP, PTFE
Tolerance ±0.1 mL
Type Single Bore Buret
Class Class B