Learn More
Azlon™ Lid for Square Shaped HDPE Bucket
Product Code. 12085562
Click to view available options
Color:
Black
White
Unit Size:
Each
Product Code. 12085562
Brand: Azlon BWX118
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use with square shaped HDPE bucket
  • Lids with moulded inset handle for square shaped buckets
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lid
Material HDPE
Autoclavable Not Autoclavable
Color White
For Use With (Equipment) Square Bucket
Height (Metric) 45 mm
Length (Metric) 280 mm
Quantity 1 Ea.
Width (Metric) 280 mm