Learn More
Azlon™ Non-Return Valve for Vacuum Lines
Designed for use in vacuum lines and fluid handling systems
Brand:  Azlon NWH905P
Additional Details : Weight : 0.03000kg
Product Code. 15708047
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Neoprene disc valve has no spring and will operate at very low back pressures (NWH905P valve is now manufactured from nitrile rubber)
  • Useful for preventing back pressure in filter flasks
  • Printed arrow indicates the direction of flow
  • Rigid with stepped tapered arms to give firm grip on tubing
  • Easily disassembled for cleaning
Specifications
Specifications
Non-return Valve
Polypropylene, Neoprene
8 to 10 mm
For preventing back pressure in filter flasks