Learn More
Azlon™ Octagonal Magnetic Stirrer Bar
For stirring applications. Azlon™ Octagonal Magnetic Stirrer Bars are available in multiple lengths.
7.76€ - 223.64€
Products 6
Product Code Brand Price Quantity & Availability  
Product Code Brand Price Quantity & Availability  
10402405
View Documents Promotion Details
Azlon
SWN716
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15718097
View Documents Promotion Details
Azlon
SWN7010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
10309804
View Documents Promotion Details
Azlon
SWN708
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
10797255
View Documents Promotion Details
Azlon
SWN710
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
10696865
View Documents Promotion Details
Azlon
SWN712
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
10115924
View Documents Promotion Details
Azlon
SWN714
Length: 51mm; O.D.: 10mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • With central pivot ring
  • Shape gives increased turbulence at low speeds