missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polycarbonate Boston Round Bottles

Product Code. 15394931
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Graduations:
50mL
100mL
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 15394931

Brand: Azlon BRC0500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for preparation and storage of biological media, reagents and aqueous solutions

  • Highly transparent polycarbonate media bottles with linerless polypropylene cap
  • Suitable for repeated sterilisation by autoclaving at 121°C
  • Leakproof
  • Classic round design
  • Heavy duty and durable in use
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Diameter (Metric) Inner Neck 28 mm
For Use With (Application) Preparation and Storage of Biological Media, Reagents and Aqueous Solutions
Includes Cap
Material Polycarbonate
Neck ID (Metric) 28mm
Quantity 12/Pk.
Transparency Yes
Graduations 50mL
Graduated Yes
Shape Round
Height (Metric) 175 mm
Product Type Boston Round Bottle
Closure Material Polypropylene
Diameter (Metric) 81 mm
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.