Learn More
Azlon™ Polystyrene Specimen Container with Polyethylene Lid
Product Code. 11363614
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
100 mL
150 mL
200 mL
250 mL
Diameter (Metric):
32 mm
36 mm
51 mm
65 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 11363614
Brand: Azlon BGG452P
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Store samples with maximum visibility. Azlon™ Polystyrene Specimen Containers with Polyethylene Lids contain snap-on lids that are very easy to use.
  • Clear polystyrene containers with maximum sample visibility
  • Polyethylene airtight snap-on lids for quick easy use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Container
Closure Included Yes
Material Polystyrene
Sterility Non-sterile
Capacity (Metric) 150 mL
Closure Type Snap Cap
Color Clear
Diameter (Metric) 65 mm
Height (Metric) 53 mm
Includes Polyethylene lid
No. per Pack 10/Pk.