Learn More
Azlon™ Screw caps for Wide Mouth Vented Wash Bottles
Catalog Number p-7116203
21.23 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Caps for wide neck wash bottles with dip-tube and patented DripLok™ vapor venting valve that prevents solvent drips.
  • Available in six colors
  • One-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Closure Material Polypropylene
Color Blue
Quantity 5/Pk.