Learn More
Synergy Healthcare™ Azo™ Flat Reel Sterilizer Bags
Click to view available options
Dimensions (L x W):
200m x 100mm
200m x 150mm
200m x 200mm
Unit Size:
Pack of 2
Pack of 4
Description
- Protects its contents from deterioration, damage and contamination
- Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | EN868-5 and ISO11607-1 |
| Closure Type | Heat Seal |
| Color | Blue |
| Disposable | Single-use |
| Material | Polyethylene |
| Sterility | Non-sterile |
| No. per Case | 4 reels |
| Dimensions (L x W) | 200m x 100mm |
| Length (Metric) | 200 m |
| Show More |