missing translation for 'onlineSavingsMsg'
Learn More

Azoxystrobin-d4, TRC

Product Code. 30339111
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30339111

Brand: Toronto Research Chemicals TRCA96515210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azoxystrobin d4
CAS 1346606-39-0
Purity Grade Notes HPLC
Molecular Formula C22 2H4 H13 N3 O5
Quantity 10 mg
Synonym Methyl 2-[2-[6-(2-cyano-3,4,5,6-tetradeuteriophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoate
InChI Formula InChI=1S/C22H17N3O5/c1-27-13-17(22(26)28-2)16-8-4-6-10-19(16)30-21-11-20(24-14-25-21)29-18-9-5-3-7-15(18)12-23/h3-11,13-14H,1-2H3/b17-13+/i3D,5D,7D,9D
SMILES [2H]c1c([2H])c([2H])c(C#N)c(Oc2cc(Oc3ccccc3\C(=C/OC)\C(=O)OC)ncn2)c1[2H]
IUPAC Name methyl (E)-2-[2-[6-(2-cyano-3,4,5,6-tetradeuteriophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoate
Molecular Weight (g/mol) 407.41
Formula Weight 407.1419
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.