missing translation for 'onlineSavingsMsg'
Learn More

Azur Eosine Methylene Blue, Chem Lab™

Product Code. 11973325
Change view
Click to view available options
Quantity:
1 L
Unit Size:
Each
Product Code. Quantity unitSize
11973325 1 L Each
1 options
This item is not returnable. View return policy

Product Code. 11973325

Brand: Chem-Lab CL04.0502.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Blue
Quantity 1 L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Azur-Eosine-Methylenblue solution according Giemsa
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : glycerol, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.