missing translation for 'onlineSavingsMsg'
Learn More
Learn More
b-Anilinoethanol, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | beta-Anilinoethanol |
| CAS | 122-98-5 |
| Molecular Formula | C8H11NO |
| Quantity | 50 g |
| Synonym | 2-(Phenylamino)ethanol, 2-Anilinoethanol, N-(2-Hydroxyethyl)benzenamine, N-(2-Hydroxyethyl)aniline, N-(2-Hydroxyethyl)phenylamine, N-(beta-Hydroxyethyl)anilineN-Anilinoethanol, N-Ethanolaniline, N-Phenyl-2-aminoethanol, N-Phenylethanolamine, NSC 8393 |
| InChI Formula | InChI=1S/C8H11NO/c10-7-6-9-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2 |
| SMILES | OCCNc1ccccc1 |
| IUPAC Name | 2-anilinoethanol |
| Molecular Weight (g/mol) | 137.18 |
| Formula Weight | 137.0841 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction