missing translation for 'onlineSavingsMsg'
Learn More

Bacteriological Peptone, Ultrapure, Thermo Scientific™

Product Code. 15895388 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Unit Size:
1lb
25lb
5lb
This item is not returnable. View return policy

Product Code. 15895388

Brand: Thermo Scientific J20048.TD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

High grade peptone prepared specifically for general bacteriological purposes.

  • Format: Powder
  • CAS: 73049-73-7
  • MDL Number: MFCD00131829
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Grade Ultrapure
Product Type Bacteriological Peptone
For Use With (Application) For General Bacteriological Purposes
Packaging Type Plastic Bottle
Quantity 1 lb.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.