Learn More
Interscience™ BagRack 3500-2
Product Code. 17140309
Click to view available options
Holds:
Bags
Unit Size:
Each
Product Code. 17140309
Brand: Interscience™ 221352
Only null left
Add to basket
Add to basket
BagRack allows for the clean and orderly storage of bags during sample preparation.
  • BagRack offers the possibility of leaving bags open in a sterile area to easily pipet their contents or store them closed with a BagClip
Specifications
Product Type Bag Rack
Autoclavable Not Autoclavable
Color Light Gray
Holds Bags
Material Stainless Steel
Positions Standing
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Application) Equipment Storage During Sample Preparation
For Use With (Equipment) Sample Bags
Size 21 x 35.4 x 20 cm
Length (Metric) 210 mm
Length (English) 8.27 in.
Width (Metric) 354 mm
Width (English) 13.94 in.
Height (Metric) 200 mm
Height (English) 7.87 in.
Weight (English) 2.45 lb.
Weight (Metric) 1.11 kg