missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAPTA AM

Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Selective calcium chelator that is the cell-permeable analog of BAPTA (Cat. No. 2786). Blocks hKv1.5, hKV11.1 (hERG) and hKv1.3 channels (Ki values are 1.23, 1.30 and 1.45 μM respectively). Displays antithrombotic activity in vitro.
F 127 (Cat. No. 6253) for the solubilization of BAPTA AM is also available.
Specifications
Specifications
| CAS | 126150-97-8 |
| Quantity | 25 mg |
| Molecular Formula | C34H40N2O18 |
| Formula Weight | 764.68 |
| Purity | >95% |
| Molecular Weight (g/mol) | 764.69 |
| Chemical Name or Material | BAPTA AM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction