Learn More
Masterflex™ Barbed Elbow/tee/Y-connector Kits, Nylon
Organize the fittings at your workbench with these convenient fittings kits
119.00€ - 140.00€
Specifications
Product Type Barbed elbow/tee/Y-connector kits, Nylon, fittings kits (set of 1)
Material Nylon
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
13413866
View Documents Promotion Details
Masterflex™
31514-14
Elbows: 1/16 in., 3/32 in., 1/8 in., 5/32 in., 3/16 in., 1/4 in., 5/16 in. Tees: 1/16 in., 3/32 in., 1/8 in., 5/32 in., 3/16 in., 1/4 in., 5/16 in. Y-connectors: 1/16 in., 3/16 in., 1/4 in., 3/8 in.
140.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
15108335
View Documents Promotion Details
Masterflex™
31514-18
Elbows: 1/4 in., 3/8 in., 1/2 in., 5/8 in., 3/4 in., 1 in. Tees: 1/4 in., 3/8 in., 1/2 in., 5/8 in., 3/4 in.
119.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
  • Designed for quick and easy assembly without tools.
  • Combine these simple and economical fittings with flexible tubing.
  • For use in low-pressure applications (generally less than 30psi).
Specifications
Specifications
Barbed elbow/tee/Y-connector kits, Nylon, fittings kits (set of 1)
Nylon