missing translation for 'onlineSavingsMsg'
Learn More

Barium 1-methoxy-2-propoxide, 25% w/w in 1-methoxy-2-propanol, Thermo Scientific™

Catalog Number p-7051461
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
130812-47-4
C8H18BaO4
315.555
MFCD00078021
WCXRGQPXPCQKSJ-UHFFFAOYSA-N
bis 2-ethoxyethoxy barium, barium methoxypropoxide w/w in methoxypropanol, barium bis 2-ethoxyethan-1-olate, barium 2+ ion bis 2-ethoxyethanolate
14619883
barium(2+);2-ethoxyethanolate
CCOCC[O-].CCOCC[O-].[Ba+2]
This item is not returnable. View return policy
130812-47-4
C8H18BaO4
315.555
MFCD00078021
WCXRGQPXPCQKSJ-UHFFFAOYSA-N
bis 2-ethoxyethoxy barium, barium methoxypropoxide w/w in methoxypropanol, barium bis 2-ethoxyethan-1-olate, barium 2+ ion bis 2-ethoxyethanolate
14619883
barium(2+);2-ethoxyethanolate
CCOCC[O-].CCOCC[O-].[Ba+2]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.04 g/mL
Flash Point 35°C (95°F)
Linear Formula Ba[OCH(CH3)CH2OCH3]2
Quantity 5 g
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 315.55
Concentration or Composition (by Analyte or Components) 25% w/w in 1-methoxy-2-propanol
Physical Form Liquid
Chemical Name or Material Barium 1-methoxy-2-propoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'