missing translation for 'onlineSavingsMsg'
Learn More

Barium zirconium isopropoxide, 10% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog Number p-7051945
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00798551
This item is not returnable. View return policy
MFCD00798551

Precursor of barium zirconium oxide using Sol-Gel method

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Packaging ChemSeal™ bottles
Quantity 25 mL
UN Number UN3274
Sensitivity Moisture Sensitive
Formula Weight 583.08
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Barium zirconium isopropoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'