Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Tienda Thermo Scientific Productos
Click to view available options
For Use With (Equipment):
E-Pure 3-holder
E-Pure 3-holder, Organic free
E-Pure 4-holder
E-Pure 4-holder, Organic free
E-Pure Ultrapure Water Purification System
Feedwater Source:
N/A
Pretreated
Pretreated feed
Tap
Tamaño de la unidad:
Each
Pack of 3
Descripción
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Especificaciones
Especificaciones
| Description | Cartridge Kit for E-Pure 4-holder; Pretreated feed, Organic free |
| Feedwater Source | Pretreated |
| For Use With (Equipment) | E-Pure 4-holder, Organic free |
| Type | Cartridge Kit |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido