Learn More
Thermo Scientific™ Barnstead™ MegaPure™ ACS
Shop All Thermo Scientific Products
Click to view available options
Type:
Automatic Collection System
Wall-mount Bracket
For Use With (Equipment):
Automatic Collection System
Mega-Pure Glass Still
Unit Size:
Each
Description
Control the operation of your glass still with the Thermo Scientific™ Barnstead™ MegaPure™ ACS (Automatic Collection System), which collects up to 45L of water.
Specifications
Specifications
| For Use With (Equipment) | Mega-Pure Glass Still |
| Type | Automatic Collection System |
| Dimensions (L x W x H) | 16 x 16 x 38 in. (41 x 41 x 97 cm) |
| Length (English) | 16 in. |
| Length (Metric) | 41 cm |
| Width (English) | 16 in. |
| Width (Metric) | 41 cm |
| Height (English) | 38 in. |
| Height (Metric) | 97 cm |